accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175137-62-9

IUPAC Name: 2-(thiophene-2-sulfonyl)acetonitrile
Synonyms: 2-(thiophene-2-sulfonyl)acetonitrile
Molecular Weight (g/mol): 187.23
Molecular Formula: C6H5NO2S2
InChi Key: JWLFPYPXODBUFB-UHFFFAOYSA-N
SMILES: O=S(=O)(CC#N)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results