accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHHN

CAS RN 1752-94-9

IUPAC Name: 4-(4,5-diphenyl-2,3-dihydro-1H-imidazol-2-ylidene)cyclohexa-2,5-dien-1-one
Synonyms: 4-(4,5-diphenyl-1,3-dihydroimidazol-2-ylidene)cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 312.37
Molecular Formula: C21H16N2O
InChi Key: DSSGEKZBBWKOCA-UHFFFAOYSA-N
SMILES: O=C1C=CC(C=C1)=C1NC(=C(N1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results