Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4,5-Diphenyl-1H-imidazol-2-yl)phenol 98.0+%, TCI America™
SDP

Catalog No. D4178100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4178100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4178100MG Supplier TCI America Supplier No. D4178100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1752-94-9
Molecular Formula C21H16N2O
Molecular Weight (g/mol) 312.372
MDL Number MFCD00740542
InChI Key DSSGEKZBBWKOCA-UHFFFAOYSA-N
Synonym 2-(4-Hydroxyphenyl)-4,5-diphenylimidazole
PubChem CID 5385986
IUPAC Name 4-(4,5-diphenyl-1,3-dihydroimidazol-2-ylidene)cyclohexa-2,5-dien-1-one
SMILES C1=CC=C(C=C1)C2=C(NC(=C3C=CC(=O)C=C3)N2)C4=CC=CC=C4

Specifications

Melting Point 258°C
Color Yellow
Quantity 100 mg
Formula Weight 312.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,5-Diphenyl-1H-imidazol-2-yl)phenol [for Biochemical Research]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.