CAS RN 175204-45-2
IUPAC Name:
4-(bromomethyl)-2,6-dichloropyridine
Synonyms:
4-(bromomethyl)-2,6-dichloropyridine
Molecular Weight (g/mol):
240.91
Molecular Formula:
C6H4BrCl2N
InChi Key:
XDKGMGJZRFOZRV-UHFFFAOYSA-N
SMILES:
ClC1=CC(CBr)=CC(Cl)=N1
1
–
1
of
1
results
1
Promotions Available
4-Bromomethyl-2,6-dichloropyridine, 97%, Thermo Scientific™
4-Bromomethyl-2,6-dichloropyridine, 97%, Thermo Scientific™
CAS: 175204-45-2 Molecular Formula: C6H4BrCl2N Molecular Weight (g/mol): 241 MDL Number: MFCD00052637 InChI Key: XDKGMGJZRFOZRV-UHFFFAOYSA-N Synonym: 4-bromomethyl-2,6-dichloropyridine,pyridine,4-bromomethyl-2,6-dichloro,4-bromomethyl-2 6-dichloropyridine,4-bromomethyl-2,6-dichloropyridin,4-bromomethyl-2,6-dichloro-pyridine,2,6-dichloro-4-bromomethyl pyridine,4-bromomethyl-2,6-bis chloranyl pyridine PubChem CID: 2781829 IUPAC Name: 4-(bromomethyl)-2,6-dichloropyridine SMILES: C1=C(C=C(N=C1Cl)Cl)CBr