Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromomethyl-2,6-dichloropyridine, 97%, Thermo Scientific™

Catalog No. AC433440010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433440010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC433440010 Supplier Thermo Scientific Chemicals Supplier No. 433440010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 175204-45-2
Molecular Formula C6H4BrCl2N
Molecular Weight (g/mol) 241
MDL Number MFCD00052637
InChI Key XDKGMGJZRFOZRV-UHFFFAOYSA-N
Synonym 4-bromomethyl-2,6-dichloropyridine, pyridine,4-bromomethyl-2,6-dichloro, 4-bromomethyl-2 6-dichloropyridine, 4-bromomethyl-2,6-dichloropyridin, 4-bromomethyl-2,6-dichloro-pyridine, 2,6-dichloro-4-bromomethyl pyridine, 4-bromomethyl-2,6-bis chloranyl pyridine
PubChem CID 2781829
IUPAC Name 4-(bromomethyl)-2,6-dichloropyridine
SMILES C1=C(C=C(N=C1Cl)Cl)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 42.5°C to 43.2°C
Color Yellow
Boiling Point 118.0°C (0.2 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 241
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromomethyl-2, 6-dichloropyridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.