accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrFFF

CAS RN 175205-82-0

IUPAC Name: 2-bromo-3-(trifluoromethyl)pyridine
Synonyms: 2-bromo-3-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 226
Molecular Formula: C6H3BrF3N
InChi Key: OFGSIPQYQUVVPL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(Br)N=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)