Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-(trifluoromethyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. B31795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31795G 5 g
B31791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31795G Supplier TCI America Supplier No. B31795G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 175205-82-0
Molecular Formula C6H3BrF3N
Molecular Weight (g/mol) 225.996
MDL Number MFCD00153084
InChI Key OFGSIPQYQUVVPL-UHFFFAOYSA-N
Synonym 2-bromo-3-trifluoromethyl pyridine, 2-bromo-3-trifluorormethyl pyridine, pyridine, 2-bromo-3-trifluoromethyl, 2-bromo-3-trifluormethyl-pyridine, pubchem2999, acmc-1bx8n, ksc174i2d, tpc-py049, buttpark 27\06-85
PubChem CID 5193820
IUPAC Name 2-bromo-3-(trifluoromethyl)pyridine
SMILES C1=CC(=C(N=C1)Br)C(F)(F)F

Specifications

Melting Point 40°C
Boiling Point 69°C
Quantity 5 g
UN Number 2811
Formula Weight 226.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.