accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NNOHOCH3

CAS RN 175277-11-9

IUPAC Name: 3-tert-butyl-1-methyl-1H-pyrazole-5-carboxylic acid
Synonyms: 5-tert-butyl-2-methylpyrazole-3-carboxylic acid
Molecular Weight (g/mol): 182.22
Molecular Formula: C9H14N2O2
InChi Key: SFSXXMXHJOSBAZ-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C(O)=O)C(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results