Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid, 95%, Thermo Scientific™

Catalog No. SPB03149EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SPB03149EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPB03149EA Supplier Thermo Scientific Chemicals Supplier No. SPB03149EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 175277-11-9
Molecular Formula C9H14N2O2
Molecular Weight (g/mol) 182.223
InChI Key SFSXXMXHJOSBAZ-UHFFFAOYSA-N
Synonym 3-tert-butyl-1-methyl-1h-pyrazole-5-carboxylic acid, 3-tert-butyl-1-methylpyrazole-5-carboxylic acid, 5-tert-butyl-2-methyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 3-1,1-dimethylethyl-1-methyl, 3-t-butyl-1-methyl-1h-pyrazole-5-carboxylic acid, maybridge1_008671, acmc-1bzn5, butylmethylpyrazolecarboxylicacid
PubChem CID 720529
IUPAC Name 5-tert-butyl-2-methylpyrazole-3-carboxylic acid
SMILES CC(C)(C)C1=NN(C(=C1)C(=O)O)C

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 182.22
Percent Purity 95%
Chemical Name or Material 3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.