accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHNHCl

CAS RN 175277-40-4

IUPAC Name: 1-(1H-pyrazol-5-yl)ethan-1-one hydrochloride
Synonyms: 1-(2H-pyrazol-3-yl)ethanone hydrochloride
Molecular Weight (g/mol): 146.57
Molecular Formula: C5H7ClN2O
InChi Key: MTFWBEDWXAHBNR-UHFFFAOYSA-N
SMILES: Cl.CC(=O)C1=CC=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results