Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1H-Pyrazol-5-yl)ethan-1-one hydrochloride, ≥95%, Thermo Scientific™

Catalog No. SPB05355EA
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPB05355EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPB05355EA Supplier Thermo Scientific Chemicals Supplier No. SPB05355EA
Add to Cart

Chemical Identifiers

CAS 175277-40-4
Molecular Formula C5H7ClN2O
Molecular Weight (g/mol) 146.574
MDL Number MFCD00204160
InChI Key MTFWBEDWXAHBNR-UHFFFAOYSA-N
Synonym 1-1h-pyrazol-5-yl ethan-1-one hydrochloride, 1-1h-pyrazol-5-yl ethanone hydrochloride, 1-1h-pyrazol-3-yl ethanone hydrochloride, 1-2h-pyrazol-3-yl ethanone hydrochloride, 5-acetyl-1h-pyrazole hydrochloride, 5-acetylpyrazole, chloride, 3-acetylpyrazole hydrochloride, 5-acetylpyrazole hydrochloride, 1-1h-pyrazol-5-yl-1-ethanone hydrochloride, 1-1h-pyrazole-5-yl ethan-1-one hydrochloride
PubChem CID 2799623
IUPAC Name 1-(1H-pyrazol-5-yl)ethanone;hydrochloride
SMILES CC(=O)C1=CC=NN1.Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 146.58
Percent Purity 95%
Chemical Name or Material 1-(1H-pyrazol-5-yl)ethan-1-one hydrochloride
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.