accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175278-23-6

IUPAC Name: 4-amino-3-(trifluoromethoxy)benzonitrile
Synonyms: 4-amino-3-(trifluoromethoxy)benzonitrile
Molecular Weight (g/mol): 202.14
Molecular Formula: C8H5F3N2O
InChi Key: QNJPSMLILRHZOW-UHFFFAOYSA-N
SMILES: NC1=C(OC(F)(F)F)C=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results