accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH3H3C

CAS RN 1754-55-8

IUPAC Name: ethyl 2,4,6-trimethylbenzoate
Synonyms: ethyl 2,4,6-trimethylbenzoate
Molecular Weight (g/mol): 192.26
Molecular Formula: C12H16O2
InChi Key: ZXTXIZPSMQCYBN-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)C=C(C)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results