Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2,4,6-Trimethylbenzoate 95.0+%, TCI America™
SDP

Catalog No. E073625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E073625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E073625G Supplier TCI America Supplier No. E073625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1754-55-8
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00015439
InChI Key ZXTXIZPSMQCYBN-UHFFFAOYSA-N
Synonym 2,4,6-Trimethylbenzoic Acid Ethyl Ester, Ethyl Mesitylenecarboxylate, Mesitylenecarboxylic Acid Ethyl Ester
PubChem CID 74465
IUPAC Name ethyl 2,4,6-trimethylbenzoate
SMILES CCOC(=O)C1=C(C=C(C=C1C)C)C

Specifications

Boiling Point 121°C
Quantity 25 g
Formula Weight 192.26
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2,4,6-Trimethylbenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.