accessibility menu, dialog, popup

UserName

Viewing profile as user:
SeBrBr

CAS RN 1755-36-8

IUPAC Name: 2,5-dibromoselenophene
Synonyms: 2,5-dibromoselenophene
Molecular Weight (g/mol): 288.84
Molecular Formula: C4H2Br2Se
InChi Key: QAYFAXYTKFYUDZ-UHFFFAOYSA-N
SMILES: BrC1=CC=C(Br)[Se]1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results