Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromoselenophene 98.0+%, TCI America™
SDP

Catalog No. D40285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40285G 5 g
D40281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40285G Supplier TCI America Supplier No. D40285G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1755-36-8
Molecular Formula C4H2Br2Se
Molecular Weight (g/mol) 288.839
InChI Key QAYFAXYTKFYUDZ-UHFFFAOYSA-N
PubChem CID 13439265
IUPAC Name 2,5-dibromoselenophene
SMILES C1=C([Se]C(=C1)Br)Br

Specifications

Color Yellow
Boiling Point 104°C
Quantity 5 g
UN Number 3440
Formula Weight 288.83
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dibromoselenophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.