accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1755-51-7

IUPAC Name: bis(2,4-dimethoxyphenyl)(2-methoxyphenyl)methanol
Synonyms: bis(2,4-dimethoxyphenyl)(2-methoxyphenyl)methanol
Molecular Weight (g/mol): 410.47
Molecular Formula: C24H26O6
InChi Key: GEPSNGQKRLULHW-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(OC)=C1)C(O)(C1=CC=CC=C1OC)C1=CC=C(OC)C=C1OC
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results