Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentamethoxy Red, TCI America™
SDP

Catalog No. P06311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P06311G Supplier TCI America Supplier No. P06311G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1755-51-7
Molecular Formula C24H26O6
Molecular Weight (g/mol) 410.466
MDL Number MFCD00070616
InChI Key GEPSNGQKRLULHW-UHFFFAOYSA-N
Synonym Bis(2,4-dimethoxyphenyl)(2-methoxyphenyl)methanol, 2,2′C,2′C′C,4,4′C-Pentamethoxytriphenylmethanol
PubChem CID 74466
IUPAC Name bis(2,4-dimethoxyphenyl)-(2-methoxyphenyl)methanol
SMILES COC1=CC(=C(C=C1)C(C2=C(C=C(C=C2)OC)OC)(C3=CC=CC=C3OC)O)OC

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 1 g
Formula Weight 410.47
Physical Form Crystalline Powder
Chemical Name or Material Pentamethoxy Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.