accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3C

CAS RN 1758-88-9

IUPAC Name: 2-ethyl-1,4-dimethylbenzene
Synonyms: 2-ethyl-1,4-dimethylbenzene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: AXIUBBVSOWPLDA-UHFFFAOYSA-N
SMILES: CCC1=CC(C)=CC=C1C
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results