Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-p-xylene 98.0+%, TCI America™
SDP

Catalog No. E02795ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02795ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E02795ML Supplier TCI America Supplier No. E02795ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity liquid reagent for laboratory research purposes with >98.0% purity determined by GC.

Chemical Identifiers

CAS 1758-88-9
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00059233
InChI Key AXIUBBVSOWPLDA-UHFFFAOYSA-N
Synonym 2-ethyl-p-xylene, 1,4-dimethyl-2-ethylbenzene, benzene, 2-ethyl-1,4-dimethyl, 2,5-dimethylethylbenzene, p-xylene, 2-ethyl, 1-ethyl-2,5-dimethylbenzene, 1,4-dimethyl-2-ethyl benzene, acmc-1c122, p-xylene, 2-ethyl-8ci
PubChem CID 15653
IUPAC Name 2-ethyl-1,4-dimethylbenzene
SMILES CCC1=C(C=CC(=C1)C)C

Specifications

Melting Point -54°C
Color Colorless
Quantity 5 mL
Physical Form Clear Liquid at 20°C
UN Number 3295
Boiling Point 187°C
Formula Weight 134.22
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material 2-Ethyl-p-xylene
EINECSNumber (3)-3427
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.