accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOFFFFFFF

CAS RN 17587-22-3

IUPAC Name: 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyloctane-3,5-dione
Synonyms: 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyloctane-3,5-dione
Molecular Weight (g/mol): 296.19
Molecular Formula: C10H11F7O2
InChi Key: SQNZLBOJCWQLGQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)CC(=O)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results