Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethyl-6,6,7,7,8,8,8-heptafluoro-3,5-octanedione 97.0+%, TCI America™
SDP

Catalog No. D17295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D17295G Supplier TCI America Supplier No. D17295G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17587-22-3
Molecular Formula C10H11F7O2
Molecular Weight (g/mol) 296.185
MDL Number MFCD00000435
InChI Key SQNZLBOJCWQLGQ-UHFFFAOYSA-N
Synonym 2,2-dimethyl-6,6,7,7,8,8,8-heptafluoro-3,5-octanedione, 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedione, 1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedione, 3,5-octanedione, 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl, 1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octadione, 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-octane-3,5-dione, 1,1,1,2,2,3,3-heptafluoro-7,7-dimethyloctane-4,6-dione, hfod, acmc-209eam, fod-h
PubChem CID 28614
IUPAC Name 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyloctane-3,5-dione
SMILES CC(C)(C)C(=O)CC(=O)C(C(C(F)(F)F)(F)F)(F)F

Specifications

Quantity 5 g
UN Number 1993
Formula Weight 296.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethyl-6,6,7,7,8,8,8-heptafluoro-3,5-octanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.