accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 176032-78-3

IUPAC Name: 3-(1H-1,2,4-triazol-1-yl)aniline
Synonyms: 3-(1,2,4-triazol-1-yl)aniline
Molecular Weight (g/mol): 160.18
Molecular Formula: C8H8N4
InChi Key: QOHJDIAWBVPZFL-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)N1C=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results