Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(1h-1,2,4-Triazol-1-yl)aniline, 90%, Thermo Scientific™

Catalog No. CC38414DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC38414DE 5 g Amber Glass Bottle
CC38414CB 250 mg Amber Glass Bottle
CC38414DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC38414DE Supplier Thermo Scientific Chemicals Supplier No. CC38414DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 176032-78-3
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
InChI Key QOHJDIAWBVPZFL-UHFFFAOYSA-N
PubChem CID 23503656
IUPAC Name 3-(1,2,4-triazol-1-yl)aniline
SMILES C1=CC(=CC(=C1)N)N2C=NC=N2

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 3-1h-1,2,4-triazol-1-yl aniline, 3-1,2,4-triazol-1-yl aniline, benzenamine,3-1h-1,2,4-triazol-1-yl, benzenamine, 3-1h-1,2,4-triazol-1-yl, 1-3-aminophenyl-1h-1,2,4-triazole, 3-1,2,4-triazolyl phenylamine, 3-1,2,4 triazol-1-yl-phenylamine, 3-1,2,4 triazol-1-yl phenylamine
Formula Weight 160.18
Percent Purity 90%
Chemical Name or Material 3-(1H-1, 2, 4-Triazol-1-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.