accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHNO(Z)

CAS RN 1761-56-4

IUPAC Name: (6Z)-6-{[(2-hydroxyphenyl)amino]methylidene}cyclohexa-2,4-dien-1-one
Synonyms: (6Z)-6-{[(2-hydroxyphenyl)amino]methylidene}cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 213.24
Molecular Formula: C13H11NO2
InChi Key: AAPPQBJWIDZNGZ-KTKRTIGZSA-N
SMILES: OC1=CC=CC=C1N\C=C1\C=CC=CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results