Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Salicylideneaminophenol 98.0+%, TCI America™
SDP

Catalog No. S03285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03285G 5 g
S032825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03285G Supplier TCI America Supplier No. S03285G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1761-56-4
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.236
MDL Number MFCD00020011
InChI Key AAPPQBJWIDZNGZ-KTKRTIGZSA-N
PubChem CID 5398322
IUPAC Name (6Z)-6-[(2-hydroxyanilino)methylidene]cyclohexa-2,4-dien-1-one
SMILES C1=CC=C(C(=C1)NC=C2C=CC=CC2=O)O

Specifications

Melting Point 187°C
Quantity 5 g
Formula Weight 213.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Salicylideneaminophenol
RTECSNumber SL5360000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.