accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17610-00-3

CH3H3CCH3OH3CH3CCH3

CAS RN 17610-00-3

IUPAC Name: 3,5-di-tert-butylbenzaldehyde
Synonyms: 3,5-di-tert-butylbenzaldehyde
Molecular Weight (g/mol): 218.34
Molecular Formula: C15H22O
InChi Key: BRUITYMDHWNCIG-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=CC(C=O)=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)