accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 176225-09-5

IUPAC Name: 4-fluoro-2-(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 4-fluoro-2-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 262.6
Molecular Formula: C7H3ClF4O2S
InChi Key: IGMYEVQPXWKFQF-UHFFFAOYSA-N
SMILES: FC1=CC(=C(C=C1)S(Cl)(=O)=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results