Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-2-(trifluoromethyl)benzenesulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. F11245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11245G 5 g
F11241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11245G Supplier TCI America Supplier No. F11245G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 176225-09-5
Molecular Formula C7H3ClF4O2S
Molecular Weight (g/mol) 262.603
MDL Number MFCD01091000
InChI Key IGMYEVQPXWKFQF-UHFFFAOYSA-N
Synonym 4-fluoro-2-trifluoromethyl benzenesulfonyl chloride, 4-fluoro-2-trifluoromethyl benzene-1-sulfonyl chloride, 4-fluoro-2-trifluoromethyl-benzenesulfonyl chloride, 4-fluoro-2-trifluoromethyl benzenesulphonyl chloride, 2-chlorosulphonyl-4-fluorobenzotrifluoride, 4-fluoro-2-trifluoromethyl-benzenesulfonylchloride, 4-fluoro-2-trifluoromethyl benzene sulphonyl chloride, pubchem4276, acmc-1c2ls, 2-chlorosulphonyl-5-fluorobenzotrifluoride
PubChem CID 2737548
IUPAC Name 4-fluoro-2-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC(=C(C=C1F)C(F)(F)F)S(=O)(=O)Cl

Specifications

Melting Point 50°C
Boiling Point 76°C
Quantity 5 g
UN Number 3261
Formula Weight 262.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzenesulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.