accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOSOOHS

CAS RN 17636-10-1

IUPAC Name: sodium 3-sulfanylpropane-1-sulfonate
Synonyms: sodium 3-sulfanylpropane-1-sulfonate
Molecular Weight (g/mol): 178.2
Molecular Formula: C3H7NaO3S2
InChi Key: FRTIVUOKBXDGPD-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)CCCS
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results