accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOON

CAS RN 17639-64-4

IUPAC Name: 1-(4-methylbenzenesulfonyl)pyrrolidine
Synonyms: 1-(4-methylbenzenesulfonyl)pyrrolidine
Molecular Weight (g/mol): 225.31
Molecular Formula: C11H15NO2S
InChi Key: KDWPQSBXEHQMSD-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N1CCCC1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results