Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(p-Toluenesulfonyl)pyrrole 99.0+%, TCI America™
SDP

Catalog No. T13865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T13865G Supplier TCI America Supplier No. T13865G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17639-64-4
Molecular Formula C11H11NO2S
Molecular Weight (g/mol) 221.274
MDL Number MFCD00145014
InChI Key OXWIEWFMRVJGNY-UHFFFAOYSA-N
Synonym 1-tosylpyrrole, 1-tosyl-1h-pyrrole, 1-p-toluenesulfonyl pyrrole, n-tosylpyrrole, 1-4-methylphenyl sulfonyl-1h-pyrrole, 1-p-tolylsulfonyl pyrrole, 1-tosyl-pyrrole, 1-4-methylphenylsulfonyl pyrrole, 1-4-methylbenzenesulfonyl pyrrole, 1h-pyrrole, 1-4-methylphenyl sulfonyl
PubChem CID 640087
IUPAC Name 1-(4-methylphenyl)sulfonylpyrrole
SMILES CC1=CC=C(C=C1)S(=O)(=O)N2C=CC=C2

Specifications

Melting Point 103°C
Color Purple-White
Quantity 5 g
Formula Weight 221.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(p-Toluenesulfonyl)pyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.