accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17640-25-4

IUPAC Name: methyl 2,2-dichloro-2-methoxyacetate
Synonyms: methyl 2,2-dichloro-2-methoxyacetate
Molecular Weight (g/mol): 172.99
Molecular Formula: C4H6Cl2O3
InChi Key: FWIGIASHGJFYOL-UHFFFAOYSA-N
SMILES: COC(=O)C(Cl)(Cl)OC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)