Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Dichloromethoxyacetate 95.0+%, TCI America™
SDP

Catalog No. A10465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10465G 5 g
A104625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10465G Supplier TCI America Supplier No. A10465G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17640-25-4
Molecular Formula C4H6Cl2O3
Molecular Weight (g/mol) 172.989
MDL Number MFCD00012202
InChI Key FWIGIASHGJFYOL-UHFFFAOYSA-N
PubChem CID 2724502
IUPAC Name methyl 2,2-dichloro-2-methoxyacetate
SMILES COC(=O)C(OC)(Cl)Cl

Specifications

Color Colorless
Boiling Point 181°C
Quantity 5 g
UN Number 3265
Synonym methyldichloromethoxyacetate, methyl2,2-dichloro-2-methoxyacetate, methoxydichloroacetic acid methyl ester, methyl 2,2-dichloro-2-methoxyacetate, methyl 2 pound not2-dichloro-2-methoxyacetate, aceticacid,2,2-dichloro-2-methoxy-,methylester, acetic acid,2,2-dichloro-2-methoxy-,methyl ester
Formula Weight 172.99
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Dichloromethoxyacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.