accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONOHOHNOO

CAS RN 176486-63-8

IUPAC Name: 1-[(tert-butoxy)carbonyl]-4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pyrrolidine-2-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)-4-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 452.51
Molecular Formula: C25H28N2O6
InChi Key: UPXRTVAIJMUAQR-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC(CC1C(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)