Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-trans-4-(Fmoc-amino)-L-proline, 97%, Thermo Scientific™

Catalog No. AAH66878MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66878MD 250 mg
AAH6687803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66878MD Supplier Thermo Scientific Chemicals Supplier No. H66878MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 176486-63-8
Molecular Formula C25H28N2O6
Molecular Weight (g/mol) 452.507
MDL Number MFCD00673783
InChI Key UPXRTVAIJMUAQR-VFNWGFHPSA-N
Synonym 2s,4r-4-9h-fluoren-9-yl methoxy carbonyl amino-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-boc-trans-4-n-fmoc-amino-l-proline, 2s,4r-n-tert-butoxycarbonyl-4-n-9-fluorenylmethoxycarbonyl aminopyrrolidine-2-carboxylic acid, 2s,4r-4-9h-fluoren-9-yl methoxy carbonylamino-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, 2s,4r-1-tert-butoxycarbonyl-4-9h-fluoren-9-ylmethoxy carbonyl amino pyrrolidine-2-carboxylic acid, ambotzbaa1783, 2s,4r-fmoc-4-amino-1-boc-pyrrolidine-2-carboxyli, 2s,4r-1-boc-4-n-fmoc-4-aminopyrrolidine-2-carboxylic acid, 2s,4r-trans-n-alpha-t-butyloxycarbonyl-4-9-fluorenylmethyloxycarbonyl amino-l-proline
PubChem CID 2756135
IUPAC Name (2S,4R)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 197°C to 201°C
Color White
Quantity 250 mg
Formula Weight 452.5
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Boc-trans-4-(Fmoc-amino)-L-proline
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.