CAS RN 176504-36-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Bisindolylmaleimide I, Hydrochloride
An enhanced water-soluble form of Bisindolylmaleimide I.