Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Bisindolylmaleimide I, Hydrochloride

Catalog No. 2032911MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-329-11MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-329-11MG Supplier MilliporeSigma™ Supplier No. 2032911MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An enhanced water-soluble form of Bisindolylmaleimide I.

An enhanced water-soluble form of Bisindolylmaleimide I (Cat. No. 203290). A reversible and ATP-competitive inhibitor of protein kinase C (Ki = 10 nM). May inhibit protein kinase A at much higher concentrations (Ki = 2μM).
TRUSTED_SUSTAINABILITY

Specifications

Color Orange-red
Target Proteins Protein kinase C
CAS 176504-36-2
Molecular Formula C25H24N4O2·HCl
Formula Weight 448.9g/mol
Solubility DMSO or H2O
Form Solid
Quantity 1 mg
Purity 95% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.