accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 176692-55-0

IUPAC Name: ethyl 4-methyl-5-oxo-4H,5H-[1,2,3]triazolo[1,5-a]quinazoline-3-carboxylate
Synonyms: ethyl 4-methyl-5-oxo-[1,2,3]triazolo[1,5-a]quinazoline-3-carboxylate
Molecular Weight (g/mol): 272.26
Molecular Formula: C13H12N4O3
InChi Key: KWIXXZHCRSDFBE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C2N(N=N1)C1=CC=CC=C1C(=O)N2C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results