Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-methyl-5-oxo-4,5-dihydro[1,2,3]triazolo[1,5-a]quinazoline-3-carboxylate, 95%, Thermo Scientific™

Catalog No. HTS12188DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
HTS12188DA 1 g Amber Glass Bottle
HTS12188CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. HTS12188DA Supplier Thermo Scientific Chemicals Supplier No. HTS12188DA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 176692-55-0
Molecular Formula C13H12N4O3
Molecular Weight (g/mol) 272.264
InChI Key KWIXXZHCRSDFBE-UHFFFAOYSA-N
Synonym ethyl 4-methyl-5-oxo-4,5-dihydro 1,2,3 triazolo 1,5-a quinazoline-3-carboxylate, ethyl 4-methyl-5-oxo-1,2,3 triazolo 1,5-a quinazoline-3-carboxylate, maybridge4_001995, ethyl 4-methyl-5-oxotriazolo 1,5-a quinazoline-3-carboxylate, 4,5-dihydro-5-oxo-4-methyl 1,2,3 triazolo 1,5-a quinazoline-3-carboxylic acid ethyl ester, ethyl 4-methyl-5-oxo-4,5-dihydro-1,2,3 triazolo 1,5-a quinazoline-3-carboxylate, ethyl4-methyl-5-oxo-4,5-dihydro 1,2,3 triazolo 1,5-a quinazoline-3-carboxylate, 1,2,3 triazolo 1,5-a quinazoline-3-carboxylicacid, 4,5-dihydro-4-methyl-5-oxo-, ethyl ester, ethyl 7-methyl-8-oxo-2,3,4,7-tetraazatricyclo 7.4.0.0^ 2,6 trideca-1 9 ,3,5,10,12-pentaene-5-carboxylate
PubChem CID 2816163
IUPAC Name ethyl 4-methyl-5-oxotriazolo[1,5-a]quinazoline-3-carboxylate
SMILES CCOC(=O)C1=C2N(C(=O)C3=CC=CC=C3N2N=N1)C

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 272.26
Chemical Name or Material Ethyl 4-methyl-5-oxo-4,5-dihydro[1,2,3]triazolo[1,5-a]quinazoline-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.