accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17681-50-4

IUPAC Name: tetrasodium (3E)-5-benzamido-3-{2-[5-({4-chloro-6-[(4-sulfonatophenyl)amino]-1,3,5-triazin-2-yl}amino)-2-sulfonatophenyl]hydrazin-1-ylidene}-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: tetrasodium (3E)-5-benzamido-3-{2-[5-({4-chloro-6-[(4-sulfonatophenyl)amino]-1,3,5-triazin-2-yl}amino)-2-sulfonatophenyl]hydrazin-1-ylidene}-4-oxonaphthalene-2,7-disulfonate
Molecular Weight (g/mol): 995.2
Molecular Formula: C32H19ClN8Na4O14S4
InChi Key: HMBFFLCYQUKBIW-FORSLLQSSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(NC2=NC(Cl)=NC(NC3=CC=C(C(N\N=C4/C(=O)C5=C(NC(=O)C6=CC=CC=C6)C=C(C=C5C=C4S([O-])(=O)=O)S([O-])(=O)=O)=C3)S([O-])(=O)=O)=N2)C=C1
Quantity 
  • (1)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results