Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reactive Red 4, MP Biomedicals™, MP Biomedicals™

Catalog No. ICN15201725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15201725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15201725 Supplier MP Biomedicals, Inc Supplier No. 0215201725
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17681-50-4
Molecular Formula C32H19ClN8Na4O14S4
Molecular Weight (g/mol) 995.195
InChI Key HMBFFLCYQUKBIW-UHFFFAOYSA-J
Synonym C.I. 18105, Cibacron Brilliant Red 3B-A
PubChem CID 44134921
IUPAC Name tetrasodium;5-benzamido-3-[[5-[[4-chloro-6-(4-sulfonatoanilino)-1,3,5-triazin-2-yl]amino]-2-sulfonatophenyl]hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)C(=O)NC2=C3C(=CC(=C2)S(=O)(=O)[O-])C=C(C(=NNC4=C(C=CC(=C4)NC5=NC(=NC(=N5)NC6=CC=C(C=C6)S(=O)(=O)[O-])Cl)S(=O)(=O)[O-])C3=O)S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Formula Weight 995.2
Color Purple to Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Reactive Red 4
Recommended Storage Store at room temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.