accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17683-09-9

IUPAC Name: 1-benzyl-3-(4-methylphenyl)triaz-1-ene
Synonyms: 1-benzyl-3-(4-methylphenyl)triaz-1-ene
Molecular Weight (g/mol): 225.3
Molecular Formula: C14H15N3
InChi Key: IWCGEJKKDAJKJE-UHFFFAOYSA-N
SMILES: CC1=CC=C(NN=NCC2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results