Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-3-p-tolyltriazene 98.0+%, TCI America™
SDP

Catalog No. B09491G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B09491G Supplier TCI America Supplier No. B09491G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17683-09-9
Molecular Formula C14H15N3
Molecular Weight (g/mol) 225.30
MDL Number MFCD00038226
InChI Key IWCGEJKKDAJKJE-UHFFFAOYSA-N
Synonym 3-p-Tolyl-1-benzyltriazene
PubChem CID 87234
IUPAC Name 1-benzyl-3-(4-methylphenyl)triaz-1-ene
SMILES CC1=CC=C(NN=NCC2=CC=CC=C2)C=C1

Specifications

Melting Point 77°C
Quantity 1 g
UN Number 1325
Formula Weight 225.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl-3-p-tolyltriazene [for Esterification]
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.