accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17688-68-5

IUPAC Name: 4-phenyl-1λ⁶-thiomorpholine-1,1-dione
Synonyms: 4-phenyl-1λ⁶-thiomorpholine-1,1-dione
Molecular Weight (g/mol): 211.28
Molecular Formula: C10H13NO2S
InChi Key: OLQHDKQJHKZUNN-UHFFFAOYSA-N
SMILES: O=S1(=O)CCN(CC1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results