Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenylthiomorpholine 1,1-Dioxide 98.0+%, TCI America™
SDP

Catalog No. P16035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16035G 5 g
P160325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16035G Supplier TCI America Supplier No. P16035G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17688-68-5
Molecular Formula C10H13NO2S
Molecular Weight (g/mol) 211.28
MDL Number MFCD00100136
InChI Key OLQHDKQJHKZUNN-UHFFFAOYSA-N
PubChem CID 87241
IUPAC Name 4-phenyl-1λ⁶-thiomorpholine-1,1-dione
SMILES O=S1(=O)CCN(CC1)C1=CC=CC=C1

Specifications

Melting Point 120°C
Color Purple-White
Quantity 5 g
Formula Weight 211.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylthiomorpholine 1,1-Dioxide
RTECSNumber XM3050025
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.