accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1769-24-0

IUPAC Name: 2-methyl-1,4-dihydroquinazolin-4-one
Synonyms: 2-methyl-4(3H)-quinazolinone
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: FIEYHAAMDAPVCH-UHFFFAOYSA-N
SMILES: CC1=NC(=O)C2=CC=CC=C2N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results