Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-4(1H)-quinazolinone 98.0+%, TCI America™
SDP

Catalog No. M27095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M27095G Supplier TCI America Supplier No. M27095G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1769-24-0
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.18
MDL Number MFCD00006887,MFCD05270928
InChI Key FIEYHAAMDAPVCH-UHFFFAOYSA-N
PubChem CID 15674
IUPAC Name 2-methyl-1,4-dihydroquinazolin-4-one
SMILES CC1=NC(=O)C2=CC=CC=C2N1

Specifications

Melting Point 240°C
Color White-Yellow
Quantity 5 g
Formula Weight 160.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-4(1H)-quinazolinone
RTECSNumber VA3677000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.