accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17696-60-5

CH3ONNOCH3(E/Z)(E/Z)

CAS RN 17696-60-5

IUPAC Name: N-(4-ethoxyphenyl)-1-(4-{[(4-ethoxyphenyl)imino]methyl}phenyl)methanimine
Synonyms: N-(4-ethoxyphenyl)-1-(4-{[(4-ethoxyphenyl)imino]methyl}phenyl)methanimine
Molecular Weight (g/mol): 372.47
Molecular Formula: C24H24N2O2
InChi Key: KSHALPAIUOUGBA-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)N=CC1=CC=C(C=NC2=CC=C(OCC)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results