accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOOH

CAS RN 17696-61-6

IUPAC Name: butan-2-yl 4-hydroxybenzoate
Synonyms: sec-butyl 4-hydroxybenzoate
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: ZUOTXZHOGPQFIU-UHFFFAOYNA-N
SMILES: CCC(C)OC(=O)C1=CC=C(O)C=C1
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)